Okadasäure Natriumsalz, Millipore®

Lieferant: Merck Millipore Calbiochem

459620-25EA 128 EUR
459620-25
Okadasäure Natriumsalz, Millipore®
Okadaic acid sodium salt
Formel: C₄₄H₆₇NaO₁₃
Molecular Weight: 827,00 g/mol
MDL: MFCD00210208
CAS-Nummer: 209266-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR