Okadasäure

Lieferant: BIO SOURCE INT

BSOUPHZ1154EA 0 EUR
BSOUPHZ1154
Okadasäure
Okadaic acid
Formel: C₄₄H₆₈O₁₃
Molecular Weight: 805,02 g/mol
MDL: MFCD00083455
CAS-Nummer: 78111-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR