4-(Chlormethyl)-3,5-dimethylisoxazol

Lieferant: CHEMPUR

CHMPJFD00628.250EA 0 EUR
CHMPJFD00628.250 MAYBJFD00628DA
4-(Chlormethyl)-3,5-dimethylisoxazol
4-(Chloromethyl)-3,5-dimethylisoxazole
Formel: C₆H₈ClNO
Molecular Weight: 145,59 g/mol
MDL: MFCD00003154
CAS-Nummer: 19788-37-5
EINECS: 243-312-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR