5-Brom-1-methyl-1H-indazol
Lieferant: COMBI-BLOCKS INC.
COBBIN-0194-5GEA
174
EUR
COBBIN-0194-5G
COBBIN-0194-1G
5-Brom-1-methyl-1H-indazol
5-Bromo-1-methyl-1H-indazole
Formel:
C₈H₇BrN₂ Molecular Weight: 211,0600 g/mol |
MDL:
MFCD09878568 CAS-Nummer: 465529-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...