1-Methyl-1H-indazol-5-boronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-9020-1GEA 251 EUR
COBBBB-9020-1G
1-Methyl-1H-indazol-5-boronsäure
1-Methyl-1H-indazole-5-boronic acid
Formel: C₈H₉BN₂O₂
MDL: MFCD09870052
CAS-Nummer: 590418-08-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR