Tri(1-naphthyl)phosphin ≥97%
|
Achtung
|
42304.04EA
95.3
EUR
42304.04
42304.18
42304.09
42304.02
Tri(1-naphthyl)phosphin ≥97%
Tri(1-naphthyl)phosphine
Formel:
C₃₀H₂₁P Molecular Weight: 412,46 g/mol Schmelzpunkt: 265…268 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046400 CAS-Nummer: 3411-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...