Triallylphosphin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL336939-5GEA 352 EUR
SIAL336939-5G
Triallylphosphin, Sigma-Aldrich®
Triallylphosphin
Formel: C₉H₁₅P
Molecular Weight: 154,189 g/mol
MDL: MFCD00014956
CAS-Nummer: 16523-89-0
EINECS: 240-591-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR