4-(4-Brom-1H-pyrazol-1-yl)-N-methylbenzolsulfonamid
Lieferant: COMBI-BLOCKS INC.
COBBHI-1537-1GEA
135
EUR
COBBHI-1537-1G
COBBHI-1537-5G
4-(4-Brom-1H-pyrazol-1-yl)-N-methylbenzolsulfonamid
4-(4-Brom-1H-pyrazol-1-yl)-N-methylbenzolsulfonamid
Formel:
C₁₀H₁₀BrN₃O₂S |
MDL:
MFCD12756447 CAS-Nummer: 1187385-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...