4-(4-Brom-1H-pyrazol-1-yl)-N-butylbenzolsulfonamid
Lieferant: COMBI-BLOCKS INC.
COBBOT-0776-1GEA
143
EUR
COBBOT-0776-1G
COBBOT-0776-5G
4-(4-Brom-1H-pyrazol-1-yl)-N-butylbenzolsulfonamid
4-(4-Brom-1H-pyrazol-1-yl)-N-butylbenzolsulfonamid
Formel:
C₁₃H₁₆BrN₃O₂S |
MDL:
MFCD13195728 CAS-Nummer: 1199773-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...