4-(4-Brom-1H-pyrazol-1-yl)-N-diisopropylbenzolsulfonamid

Lieferant: COMBI-BLOCKS INC.

COBBOT-0780-5GEA 291 EUR
COBBOT-0780-5G COBBOT-0780-1G
4-(4-Brom-1H-pyrazol-1-yl)-N-diisopropylbenzolsulfonamid
4-(4-Brom-1H-pyrazol-1-yl)-N-diisopropylbenzolsulfonamid
Formel: C₁₅H₂₀BrN₃O₂S
MDL: MFCD12756458
CAS-Nummer: 1187386-27-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR