3-(5-Methyl-1,2,4-oxadiazol-3-yl)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-6058-250MEA 543 EUR
COBBBB-6058-250M
3-(5-Methyl-1,2,4-oxadiazol-3-yl)phenylboronsäure
3-(5-Methyl-1,2,4-oxadiazol-3-yl)phenylboronsäure
Formel: C₉H₉BN₂O₃
MDL: MFCD13195651
CAS-Nummer: 1217501-31-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR