1-Boc-piperidin-4-ylidenessigsäure

Lieferant: COMBI-BLOCKS INC.

COBBHC-2815-1GEA 53.5 EUR
COBBHC-2815-1G COBBHC-2815-5G
1-Boc-piperidin-4-ylidenessigsäure
1-Boc-piperidin-4-ylidenessigsäure
Formel: C₁₂H₁₉NO₄
MDL: MFCD04115019
CAS-Nummer: 193085-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR