2-(4-Methylcyclohexyliden)essigsäure

Lieferant: COMBI-BLOCKS INC.

COBBHC-2814-1GEA 89.5 EUR
COBBHC-2814-1G COBBHC-2814-5G
2-(4-Methylcyclohexyliden)essigsäure
2-(4-Methylcyclohexyliden)essigsäure
Formel: C₉H₁₄O₂
MDL: MFCD00029539
CAS-Nummer: 77842-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR