Dicyclopentylzink 0.4 mol/l in Diethylether, Sigma-Aldrich®

Lieferant: Merck

SIAL718025-10MLEA 125 EUR
SIAL718025-10ML SIAL718025-25ML
Dicyclopentylzink 0.4 mol/l in Diethylether, Sigma-Aldrich®
Dicyclopentylzinc
Formel: C₁₀H₁₈Zn
CAS-Nummer: 20525-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR