Ethyl-8-brom-5-methylimidazo[1,2-a]pyridin-2-carboxylat
Lieferant: COMBI-BLOCKS INC.
COBBPY-7113-1GEA
291
EUR
COBBPY-7113-1G
Ethyl-8-brom-5-methylimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-8-brom-5-methylimidazo[1,2-a]pyridin-2-carboxylat
Formel:
C₁₁H₁₁BrN₂O₂ Molecular Weight: 283,1212 g/mol |
MDL:
MFCD09475853 CAS-Nummer: 135995-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...