6-Brom-7-methylimidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: COMBI-BLOCKS INC.

COBBHC-6740-1GEA 409 EUR
COBBHC-6740-1G
6-Brom-7-methylimidazo[1,2-a]pyridin-3-carbaldehyd
6-Brom-7-methylimidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₉H₇BrN₂O
MDL: MFCD11504923
CAS-Nummer: 1072944-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR