6-Chlorimidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: COMBI-BLOCKS INC.

COBBHC-6661-1GEA 157 EUR
COBBHC-6661-1G COBBHC-6661-250M
6-Chlorimidazo[1,2-a]pyridin-3-carbaldehyd
6-Chlorimidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₈H₅ClN₂O
MDL: MFCD09909649
CAS-Nummer: 29096-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR