1,1'-Bis(diisopropylphosphino)ferrocen 98%
|
Achtung
|
ACRO363875000EA
51.5
EUR
ACRO363875000
ACRO363870025
1,1'-Bis(diisopropylphosphino)ferrocen 98%
1,1'-Bis(diisopropylphosphino)ferrocen
Formel:
C₂₂H₂₈FeP₂ Molecular Weight: 410,25 g/mol Schmelzpunkt: 53…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02684559 CAS-Nummer: 97239-80-0 |
Spezifikation Testergebnisse
Appearance | Yellow to orange Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 49 to 53 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...