5-Benzyl-3-(4-nitrophenyl)-1,2,4-oxadiazol
Lieferant: COMBI-BLOCKS INC.
COBBHC-6617-1GEA
94.5
EUR
COBBHC-6617-1G
COBBHC-6617-5G
5-Benzyl-3-(4-nitrophenyl)-1,2,4-oxadiazol
5-Benzyl-3-(4-nitrophenyl)-1,2,4-oxadiazol
Formel:
C₁₅H₁₁N₃O₃ |
MDL:
MFCD03142503 CAS-Nummer: 431908-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...