5-tert-Butyl-3-(4-nitrophenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6534-5GEA 409 EUR
COBBHC-6534-5G COBBHC-6534-1G
5-tert-Butyl-3-(4-nitrophenyl)-1,2,4-oxadiazol
5-tert-Butyl-3-(4-nitrophenyl)-1,2,4-oxadiazol
Formel: C₁₂H₁₃N₃O₃
MDL: MFCD02310127
CAS-Nummer: 1004398-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR