3-(4-Nitrophenyl)-5-propyl-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6429-1GEA 143 EUR
COBBHC-6429-1G COBBHC-6429-5G
3-(4-Nitrophenyl)-5-propyl-1,2,4-oxadiazol
3-(4-Nitrophenyl)-5-propyl-1,2,4-oxadiazole
Formel: C₁₁H₁₁N₃O₃
MDL: MFCD09972144
CAS-Nummer: 10364-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR