Ethyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat 95%

Achtung

H35022.03EA 176.8 EUR
H35022.03
Ethyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat 95%
Ethyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat
Formel: C₁₀H₉BrN₂O₂
Molecular Weight: 269,10 g/mol
Schmelzpunkt: 95…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12827886
CAS-Nummer: 1215504-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR