3-(3,4-Difluorphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-5174-1GEA 94.5 EUR
COBBBB-5174-1G COBBBB-5174-5G
3-(3,4-Difluorphenylcarbamoyl)phenylboronsäure
3-(3,4-Difluorphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₀BF₂NO₃
MDL: MFCD09972116
CAS-Nummer: 1072946-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR