2-Fluor-5-(isopropylcarbamoyl)phenylboronsäure ≥98%

Achtung

H52605.MDEA 77 EUR
H52605.MD H52605.03
2-Fluor-5-(isopropylcarbamoyl)phenylboronsäure ≥98%
2-Fluor-5-(isopropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₃BFNO₃
Molecular Weight: 225,03 g/mol
Schmelzpunkt: 130…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08235103
CAS-Nummer: 874289-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR