4-(3-Chlor-4-methylphenylcarbamoyl)phenylboronsäure ≥98%

Achtung

H53015.MDEA 66.3 EUR
H53015.MD
4-(3-Chlor-4-methylphenylcarbamoyl)phenylboronsäure ≥98%
4-(3-Chlor-4-methylphenylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₃BClNO₃
Molecular Weight: 289,53 g/mol
Schmelzpunkt: 217…219 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027211
CAS-Nummer: 913835-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR