Cyclopentylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-2074-1GEA 21.2 EUR
COBBBB-2074-1G COBBBB-2074-25G COBBBB-2074-5G
Cyclopentylboronsäure
Cyclopentylboronic acid
Formel: C₅H₁₁BO₂
Molecular Weight: 113,95 g/mol
MDL: MFCD01074541
CAS-Nummer: 63076-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR