2-Methyl-5-nitrophenylboronsäure ≥96%

Lieferant: Thermo Scientific
Achtung

H53160.MDEA 142 EUR
H53160.MD
2-Methyl-5-nitrophenylboronsäure ≥96%
2-Methyl-5-nitrophenylboronsäure
Formel: C₇H₈BNO₄
Molecular Weight: 180,96
Schmelzpunkt: 62…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00757435
CAS-Nummer: 100960-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR