3-Brom-N-ethylbenzolsulfonamid
Lieferant: COMBI-BLOCKS INC.
COBBOT-0141-1GEA
185
EUR
COBBOT-0141-1G
COBBOT-0141-5G
3-Brom-N-ethylbenzolsulfonamid
3-Brom-N-ethylbenzolsulfonamid
Formel:
C₈H₁₀BrNO₂S |
MDL:
MFCD07363819 CAS-Nummer: 871269-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...