2-Brom-N-cyclopentylbenzamid
Lieferant: COMBI-BLOCKS INC.
COBBCS-9414-1GEA
94.5
EUR
COBBCS-9414-1G
COBBCS-9414-5G
2-Brom-N-cyclopentylbenzamid
2-Brom-N-cyclopentylbenzamid
Formel:
C₁₂H₁₄BrNO |
MDL:
MFCD00751459 CAS-Nummer: 326899-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...