3'-Brom-2'-nitroacetanilid

Lieferant: COMBI-BLOCKS INC.

COBBAN-1431-1GEA 255 EUR
COBBAN-1431-1G COBBAN-1431-5G
3'-Brom-2'-nitroacetanilid
3'-Brom-2'-nitroacetanilid
Formel: C₈H₇BrN₂O₃
MDL: MFCD11855831
CAS-Nummer: 99233-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR