DyNAmo™ Flash Probe qPCR Kits
Lieferant: Thermo Fisher Scientific
F-455XLEA 2355 EUR
FINRF-455XL FINRF-455S FINRF-455L
DyNAmo™ Flash Probe qPCR Kits
  • Extremely fast protocols: combined annealing and extension step of only 15 seconds
  • Specific and sensitive detection of a wide range of template concentration
  • Included dUTP allows the use of UNG for prevention of carry-over contamination
  • Convenient 2X master mix and optimised protocols
  • Validated with all major real time instruments
  • Licenced for qPCR
DyNAmo™ Flash Probe qPCR Kit is the ideal choice for fast probe-based qPCR. The optimised reagents and short cycling times deliver maximum speed thus increasing sample throughput. Significant time savings are achieved with both conventional and fast real time PCR instruments. Performance of the DyNAmo™ Flash Probe qPCR Kit is based on an efficient Hot Start Thermus brockianus DNA polymerase. Sensitivity and the excellent amplification efficiency across a wide range of template concentrations guarantee reproducible results. DyNAmo™ Flash Probe qPCR Kit is optimised for hydrolysis probes (e.g. TaqMan probes) and it can be used with both block- and capillary-based real time PCR instruments.
Lieferumfang: All DyNAmo™ Flash qPCR probe-based kits include 2X master mix (containing Hot Start version of a modified Tbr DNA polymerase, optimised PCR buffer, 5 mM MgCl₂, dNTP mix including dUTP) and 50X ROX passive reference dye.