Jars with screw cap
Lieferant: Semadeni
SEMA2075EA 2.1 EUR
SEMA2075
Jars with screw cap
Jars, made of PP, white, with screw cap.
  • Excellent chemical resistance
  • Temperature resistance upto 130 °C
  • Easy to clean