Intensifying screens, BioMax® TranScreen LE
Lieferant: Carestream Health
Kodak® BioMax®
KODK8660789EA 1340 EUR
KODK8660789
Intensifying screens, BioMax® TranScreen LE
BioMax™ TranScreen LE (low energy) is the first intensifying screen designed to be used with medium and low energy beta isotopes i.e. ³⁵S, ³³P, ¹⁴C, ⁴⁵Ca, ⁵⁹Fe, and ³H. The BioMax™ TranScreen LE can achieve a 0,301 Ø (medium) image 5 to 35 times faster than direct autoradiography for detection of low energy radioisotopes. The BioMax™ TranScreen LE also offers better sensitivity and resolution than fluorography, eliminating toxic fluorochromes.