5-Fluor-2-(trifluormethoxy)benzylalkohol ≥97%
|
Achtung
|
H32925.MDEA
40
EUR
H32925.MD
H32925.03
5-Fluor-2-(trifluormethoxy)benzylalkohol ≥97%
5-Fluor-2-(trifluormethoxy)benzylalkohol
Formel:
C₈H₆F₄O₂ Molecular Weight: 210,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519336 CAS-Nummer: 1092460-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...