2'-Isothiocyanatoacetophenon

Lieferant: CHEMPUR

CHMPFL19472.1EA 0 EUR
CHMPFL19472.1
2'-Isothiocyanatoacetophenon
2'-Isothiocyanatoacetophenon
Formel: C₉H₇NOS
CAS-Nummer: 134470-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR