3-Phenylpropylisothiocyanat

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO009566-25GEA 0 EUR
FLUO009566-25G FLUO009566-1G FLUO009566-5G
3-Phenylpropylisothiocyanat
3-Phenylpropylisothiocyanat
Formel: C₁₀H₁₁NS
MDL: MFCD00041134
CAS-Nummer: 2627-27-2
EINECS: 220-094-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR