3-Pentylisothiocyanat

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO009559-1GEA 36.2 EUR
FLUO009559-1G FLUO009559-5G
3-Pentylisothiocyanat
3-Pentylisothiocyanat
Formel: C₆H₁₁NS
MDL: MFCD00060395
CAS-Nummer: 201224-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR