6-Isochinolinboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBFA-2458-5GEA 1220 EUR
COBBFA-2458-5G COBBFA-2458-1G
6-Isochinolinboronsäure
6-Isoquinolineboronic acid
Formel: C₉H₈BNO₂
MDL: MFCD10697418
CAS-Nummer: 899438-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR