4-Isochinolinboronsäure
Lieferant: COMBI-BLOCKS INC.
COBBBB-2436-5GEA
316
EUR
COBBBB-2436-5G
COBBBB-2436-1G
COBBBB-2436-25G
4-Isochinolinboronsäure
4-Isoquinolineboronic acid
Formel:
C₉H₈BNO₂ Molecular Weight: 172,9763 g/mol |
MDL:
MFCD03412093 CAS-Nummer: 192182-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...