2-Methoxy-3-(trifluormethyl)benzylalkohol ≥99%

Lieferant: Thermo Scientific
Achtung

H32404.06EA 337 EUR
H32404.06 H32404.03
2-Methoxy-3-(trifluormethyl)benzylalkohol ≥99%
2-Methoxy-3-(trifluormethyl)benzylalkohol
Formel: C₉H₉F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832314
CAS-Nummer: 1017778-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR