Methyl-3,5-dimethylbenzoat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL370215-5GEA 56 EUR
SIAL370215-5G
Methyl-3,5-dimethylbenzoat, Sigma-Aldrich®
Methyl-3,5-dimethylbenzoat
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,21 g/mol
MDL: MFCD00077971
CAS-Nummer: 25081-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR