Methyl-4-methyl-3-nitrobenzoat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL632171-5GEA
47.3
EUR
SIAL632171-5G
Methyl-4-methyl-3-nitrobenzoat, Sigma-Aldrich®
Methyl-4-methyl-3-nitrobenzoat
Formel:
C₉H₉NO₄ Molecular Weight: 195,18 g/mol |
MDL:
MFCD00130054 CAS-Nummer: 7356-11-8 EINECS: 230-886-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...