Methyl-3-methyl-2-nitrobenzoat

Lieferant: CHEMPUR

CHMPRH02039.1EA 0 EUR
CHMPRH02039.1 CHMPRH02039.250
Methyl-3-methyl-2-nitrobenzoat
Methyl-3-methyl-2-nitrobenzoat
Formel: C₉H₉NO₄
Molecular Weight: 195,17 g/mol
MDL: MFCD01003947
CAS-Nummer: 5471-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR