2-(Methylsulfonyl)benzoesäure
FLUO023354.1G
FLUO023354.5G
2-(Methylsulfonyl)benzoesäure
2-(Methylsulphonyl)benzoic acid
Formel:
C₈H₈O₄S Molecular Weight: 200,21 g/mol |
MDL:
MFCD00216492 CAS-Nummer: 33963-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...