4-Allyl-3-thiosemicarbazid

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO018164-10GEA 155 EUR
FLUO018164-10G
4-Allyl-3-thiosemicarbazid
4-Allyl-3-thiosemicarbazid
Formel: C₄H₉N₃S
MDL: MFCD00007619
CAS-Nummer: 3766-55-0
EINECS: 223-186-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR