4,4-Dimethyl-3-thiosemicarbazid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL357707-1GEA
56.2
EUR
SIAL357707-1G
4,4-Dimethyl-3-thiosemicarbazid, Sigma-Aldrich®
4,4-Dimethyl-3-thiosemicarbazid
Formel:
C₃H₉N₃S Molecular Weight: 119,1887 g/mol |
MDL:
MFCD00041308 CAS-Nummer: 6926-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...