5-(Trifluormethyl)chinolin-8-boronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-5393-500MEA 454 EUR
COBBBB-5393-500M COBBBB-5393-100M COBBBB-5393-1G
5-(Trifluormethyl)chinolin-8-boronsäure
5-(Trifluormethyl)chinolin-8-boronsäure
Formel: C₁₀H₇BF₃NO₂
MDL: MFCD11504856
CAS-Nummer: 1072951-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR