1-Isobutyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-8859-1GEA 118 EUR
COBBPN-8859-1G
1-Isobutyl-1H-pyrazol-4-boronsäurepinakolester
1-Isobutyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₃H₂₃BN₂O₂
Molecular Weight: 250,1449 g/mol
MDL: MFCD05663857
CAS-Nummer: 827614-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR