2-Chlor-4,5-dimethoxyanilin

Lieferant: SIA ENAMINE

ENAMEN300-25928.1GEA 417 EUR
ENAMEN300-25928.1G
2-Chlor-4,5-dimethoxyanilin
2-Chlor-4,5-dimethoxyanilin
Formel: C₈H₁₀ClNO₂
MDL: MFCD08691143
CAS-Nummer: 32829-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR