2-Chlor-4,5-dimethoxyanilin
Lieferant: COMBI-BLOCKS INC.
COBBST-2154-1GEA
945
EUR
COBBST-2154-1G
2-Chlor-4,5-dimethoxyanilin
2-Chlor-4,5-dimethoxyanilin
Formel:
C₈H₁₀ClNO₂ |
MDL:
MFCD08691143 CAS-Nummer: 32829-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...